Sodium hexafluorophosphate, anhydrous 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC6510-100G
77417-416EA 131.42 USD
77417-416
Sodium hexafluorophosphate, anhydrous 98%
Sodium hexafluorophosphate
Formula: F₆NaP
MW: 167.95 g/mol
Melting Pt: >200 °C
Density: 2.369 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011122
CAS Number: 21324-39-0
EINECS: 244-333-1
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR