Silver trifluoromethanesulfonate 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Silver trifluoromethanesulfonate, Trifluoromethanesulphonic acid silver salt, Trifluoromethanesulfonic acid silver salt

PC6450-100G PC6450-25G PC6450-5G
77417-398EA 211.5 USD
77417-398 77417-400 77417-402
Silver trifluoromethanesulfonate 98%
Silver trifluoromethanesulphonate

Increases leaving group ability of halogens; can be used for conversion of alkyl halides to triflates & acyl halides to acyl triflates; soluble in ether and other organic solvents.

Formula: CAgF₃O₃S
MW: 256.94 g/mol
Melting Pt: 356
Storage Temperature: Ambient
MDL Number: MFCD00013226
CAS Number: 2923-28-6
EINECS: 220-882-2
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR