Silver(I) hexafluorophosphate 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Silver hexafluorophosphate

PC6400-100G PC6400-10G PC6400-25G
77417-370EA 768.18 USD
77417-370 77417-372 77417-374
Silver(I) hexafluorophosphate 98%
Silver (I) hexafluorophosphate
Formula: AgF₆P
Melting Pt: 102 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003415
CAS Number: 26042-63-7
EINECS: 247-428-6
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR