Potassium fluoride, anhydrous 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC6267S-100G PC6267S-250G PC6267S-500G
77417-344EA 88.64 USD
77417-344 77417-346 77417-348
Potassium fluoride, anhydrous 99%
Potassium fluoride
Formula: KF
Boiling Pt: 1505 °C
Melting Pt: 858 °C
Density: 2.48 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR