1-Fluorooctane 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Octyl fluoride
PC5360-25G
PC5360-5G
77416-716EA
40.17
USD
77416-716
77416-718
77416-720
1-Fluorooctane 98%
1-Fluorooctane
Formula:
C₈H₁₇F MW: 132.22 Boiling Pt: 130 Density: 0.814 g/cm³ (25 °C) Flash Pt: 42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013582 CAS Number: 463-11-6 EINECS: 207-333-2 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...