1-Fluorooctane 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Octyl fluoride

PC5360-25G PC5360-5G
77416-716EA 40.17 USD
77416-716 77416-718 77416-720
1-Fluorooctane 98%
1-Fluorooctane
Formula: C₈H₁₇F
MW: 132.22
Boiling Pt: 130
Density: 0.814 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00013582
CAS Number: 463-11-6
EINECS: 207-333-2
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR