Lithium tetrafluoroborate, anhydrous 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC5010-25G
77416-036EA 21.34 USD
77416-036
Lithium tetrafluoroborate, anhydrous 98%
Lithium tetrafluoroborate
Formula: BF₄Li
MW: 93.74 g/mol
Melting Pt: 309…311 °C
Storage Temperature: Ambient
MDL Number: MFCD00011087
CAS Number: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR