Dihydrogen hexafluorozirconate 45% (w/v) in aqueous solution

Supplier: APOLLO SCIENTIFIC

PC4812-1G
77415-726EA 28.52 USD
77415-726
Dihydrogen hexafluorozirconate 45% (w/v) in aqueous solution
Dihydrogen hexafluorozirconate
Formula: H₂ZrF₆
Storage Temperature: Ambient
MDL Number: MFCD00082965
CAS Number: 12021-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR