1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Perfluoropropoxyethylene, (Heptafluoropropoxy)trifluoroethene, Perfluoro(propyl vnyl ether)

PC4513L-100G PC4513L-25G
77415-600EA 391.22 USD
77415-600 77415-602
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane 97%
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane

Useful intermediate and monomer; the co-polymer with tetrafluoroethylene is sold as Teflon PFA.

Formula: C₅F₁₀O
Boiling Pt: 35…36 °C
Melting Pt: –70 °C
Density: 1.53 g/cm³ (25 °C)
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191577
CAS Number: 1623-05-8
EINECS: 216-600-2
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR