4-Fluorophenyl isocyanate 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
4-Fluorophenylisocyanate, 1-Fluoro-4-isocyanatobenzene
PC4166-100G
PC4166-25G
PC4166-5G
77415-182EA
120.22
USD
77415-182
77415-184
77415-186
4-Fluorophenyl isocyanate 98%
4-Fluorophenyl isocyanate
- May form small amount of sediment.
Formula:
C₇H₄FNO MW: 137.11 g/mol Boiling Pt: 55 °C/8 mmHg Density: 1.206 g/cm³ (25 °C) Flash Pt: 52 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002023 CAS Number: 1195-45-5 EINECS: 214-799-0 UN: 3080 ADR: 6.1+3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...