4-Fluoro-3-phenoxybenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC4157-250G PC4157-50G PC4157-5G
77415-168EA 434.66 USD
77415-168 77415-170 77415-172
4-Fluoro-3-phenoxybenzaldehyde 98%
4-Fluoro-3-phenoxybenzaldehyde
Formula: C₁₃H₉FO₂
MW: 216.21 g/mol
Boiling Pt: 310 °C
Density: 1.209 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01318148
CAS Number: 68359-57-9
EINECS: 269-856-2
UN: 3082
ADR: 9+EHS III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR