5-Fluoro-2'-deoxyuridine (floxuridine) 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine

PC3740-1G PC3740-25G PC3740-5G
77414-862EA 32.6 USD
77414-862 77414-864 77414-866
5-Fluoro-2'-deoxyuridine (floxuridine) 99%
5-Fluoro-2'-deoxyuridine (Floxuridine)
Formula: C₉H₁₁FN₂O₅
Melting Pt: 148…151 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006530
CAS Number: 50-91-9
EINECS: 200-072-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR