4-Fluorobenzyl mercaptan 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC3721R-25G PC3721R-5G PC3721R-100G
77414-834EA 150.28 USD
77414-834 77414-836 77414-832
4-Fluorobenzyl mercaptan 98%
4-Fluorobenzyl mercaptan
Formula: C₇H₇FS
Boiling Pt: 72…74 °C/15 mmHg
Density: 1.157 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040724
CAS Number: 15894-04-9
UN: 3334
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR