(Diethylamino)sulfur trifluoride 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: DAST

PC2563-100G PC2563-25G PC2563-5G
77413-696EA 199.7 USD
77413-696 77413-698 77413-700
(Diethylamino)sulfur trifluoride 95%
(Diethylamino)sulphur trifluoride

Versatile fluorinating agent: JOC., 1975, 40, 574.


Caution: Has been reported to undergo explosive decomposition when heated above 90 °C.

Formula: C₄H₁₀F₃NS
Boiling Pt: 30…32 °C/3 mmHg
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000363
CAS Number: 38078-09-0
EINECS: 253-771-2
UN: 2920
ADR: 8+3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR