Copper(II) fluoride, anhydrous 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Cupric fluoride

PC2100-100G PC2100-25G PC2100-5G
76617-832EA 357.56 USD
76617-832 76617-834 77413-530
Copper(II) fluoride, anhydrous 99%
Copper (II) fluoride
Formula: CuF₂
MW: 101.54
Boiling Pt: 950
Melting Pt: 855 (dec.)
Density: 4.23 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010977
CAS Number: 7789-19-7
EINECS: 232-147-3
UN: 3260
ADR: 8 III
Merck Index: 12,02704

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR