Chloropentafluorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Pentafluorochlorobenzene

PC2010-5G
77413-462EA 21.42 USD
77413-462
Chloropentafluorobenzene 99%
Chloropentafluorobenzene

see: J. Organomet. Chem., 17, 145, (1969)preparation of pentafluorophenyl Grignard (Mg in THF at -10C).

Formula: C₆ClF₅
Boiling Pt: 122…123 °C/750 mmHg
Density: 1.568 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000534
CAS Number: 344-07-0
EINECS: 206-450-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR