tert-Butyldimethylsilyl trifluoromethanesulfonate 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1605-5G PC1605-100G PC1605-25G
77413-274EA 21.32 USD
77413-274 77413-270 77413-272
tert-Butyldimethylsilyl trifluoromethanesulfonate 98%
tert-Butyldimethylsilyl trifluoromethanesulphonate

Electrophilic reagent for introduction of t-butyldimethylsilyl group. For a review see: Emde et al., Synthesis, 1982 , 1.

Formula: C₇H₁₅F₃O₃SSi
Boiling Pt: 65…67 °C/12 mmHg
Density: 1.151 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000405
CAS Number: 69739-34-0
EINECS: 274-102-0
UN: 2920
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR