Bismuth(III) fluoride 99,5%

Supplier: APOLLO SCIENTIFIC
Danger

PC1230G-1G
77413-098EA 21.26 USD
77413-098
Bismuth(III) fluoride 99,5%
Bismuth (III) fluoride
Formula: BiF₃
Melting Pt: 727 °C
Density: 5.32 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010893
CAS Number: 7787-61-3
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR