2,4,6-Trifluoroanisole 98%

Supplier: APOLLO SCIENTIFIC

PC0511-10G PC0511-1G PC0511-5G
77412-596EA 158.94 USD
77412-596 77412-598 77412-600
2,4,6-Trifluoroanisole 98%
2,4,6-Trifluoroanisole
Formula: C₇H₅F₃O
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD01631502
CAS Number: 219998-30-8
UN: 3271
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR