1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Vinyl-7-oxabicyclo[4.1.0]heptane, 4-Vinyl-1,2-epoxycyclohexane

OR939803-25G
77412-076EA 20.26 USD
77412-076
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) 95%
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers)
Formula: C₈H₁₂O
Boiling Pt: 159 °C
Melting Pt: –100 °C
Density: 0.96 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022356
CAS Number: 106-86-5
UN: 3271
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR