Triethylamine 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (Diethylamino)ethane, TEA

OR9284-1L OR9284-500ML
77411-916EA 24.46 USD
77411-916 77411-918
Triethylamine 99%
Triethylamine
Formula: (C₂H₅)₃N
Boiling Pt: 89…90 °C
Melting Pt: –115 °C
Density: 0.726 g/cm³ (25 °C)
Flash Pt: –6 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR