2-Pyrrolidone

Supplier: APOLLO SCIENTIFIC
Warning

OR922249-5G OR922249-100G OR922249-25G
77411-794EA 21.36 USD
77411-794 77411-790 77411-792
2-Pyrrolidone
2-Pyrrolidone
Formula: C₄H₇NO
Boiling Pt: 245 °C
Melting Pt: 23…25 °C
Flash Pt: 113 °C
MDL Number: MFCD00005270
CAS Number: 616-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR