5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine
Supplier: APOLLO SCIENTIFIC
OR904269-1G
OR904269-25G
OR904269-5G
77411-546EA
24.46
USD
77411-546
77411-548
77411-550
5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine
5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine
Formula:
C₄H₇N₃OS Storage Temperature: Refrigerator |
MDL Number:
MFCD01203002 CAS Number: 15884-86-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...