4-Hydroxythiophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR72360-100G OR72360-25G OR72360-5G
77411-182EA 131.42 USD
77411-182 77411-184 77411-186
4-Hydroxythiophenol 98%
4-Hydroxythiophenol
Formula: C₆H₆OS
Boiling Pt: 149-150 °C
Melting Pt: 33-35 °C
Flash Pt: 110 °C
MDL Number: MFCD00004850
CAS Number: 637-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR