2-(Diphenylphosphino)biphenyl

Supplier: APOLLO SCIENTIFIC
Warning

OR72340-5G OR72340-1G OR72340-25G
77411-174EA 27.5 USD
77411-174 77411-170 77411-172
2-(Diphenylphosphino)biphenyl
2-(Diphenylphosphino)biphenyl
Formula: C₂₄H₁₉P
Melting Pt: 82 °C
MDL Number: MFCD11559063
CAS Number: 13885-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR