N,N,N',N'-Tetramethylethylenediamine (TEMED) 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR7140-250ML OR7140-500ML OR7140-100ML
77411-054EA 63.02 USD
77411-054 77411-056 77411-052
N,N,N',N'-Tetramethylethylenediamine (TEMED) 97%
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Formula: C₆H₁₆N₂
Boiling Pt: 120…122 °C
Melting Pt: –55 °C
Density: 0.775 g/cm³ (25 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR