3-Mercapto-1,2-propanediol 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR61438-10ML
77410-748EA 21.4 USD
77410-748
3-Mercapto-1,2-propanediol 97%
3-Mercapto-1,2-propanediol
Formula: C₃H₈O₂S
Boiling Pt: 118 °C/5 mmHg
Density: 1.247 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004879
CAS Number: 96-27-5
EINECS: 202-495-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR