1,4-Diisocyanatobutane

Supplier: APOLLO SCIENTIFIC
Warning

OR55710-10G OR55710-1G OR55710-5G
77410-510EA 126.34 USD
77410-510 77410-512 77410-514
1,4-Diisocyanatobutane
1,4-Diisocyanatobutane

  • May form small amount of sediment.

Formula: C₆H₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00075559
CAS Number: 4538-37-8
UN: 3334
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR