N,N,N',N',N''-Pentamethyldiethylenetriamine

Supplier: APOLLO SCIENTIFIC
Danger

OR55651-100G OR55651-500G
77410-494EA 21.4 USD
77410-494 77410-496
N,N,N',N',N''-Pentamethyldiethylenetriamine
N,N,N',N',N''-Pentamethyldiethylenetriamine
Formula: C₉H₂₃N₃
Boiling Pt: 198 °C
Melting Pt: –20 °C
Density: 0.83 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00014876
CAS Number: 3030-47-5
UN: 2922
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR