2-Amino-3-methylpyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR5258-100G OR5258-25G OR5258-500G
77410-256EA 30.56 USD
77410-256 77410-258 77410-260
2-Amino-3-methylpyridine 97%
2-Amino-3-methylpyridine
Formula: C₆H₈N₂
Boiling Pt: 221…222 °C
Melting Pt: 29…32 °C
Density: 1.073 g/cm³ (25 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00006320
CAS Number: 1603-40-3
EINECS: 216-501-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR