EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)

Supplier: APOLLO SCIENTIFIC
Danger

OR5163-100G OR5163-1KG OR5163-500G
77410-160EA 24.46 USD
77410-160 77410-162 77410-164
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride
Formula: C₈H₁₇N₃·HCl
Melting Pt: 110…115 °C
Storage Temperature: Freezer
MDL Number: MFCD00012503
CAS Number: 25952-53-8
EINECS: 247-361-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR