Triethylsilane 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR46515-250G
77409-944EA 130.92 USD
77409-944
Triethylsilane 98%
Triethylsilane
Formula: C₆H₁₆Si
Boiling Pt: 107…108 °C
Melting Pt: –157 °C
Density: 0.728 g/cm³ (25 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009018
CAS Number: 617-86-7
EINECS: 210-535-3
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR