Tricyclohexylphosphine 97%

Supplier: APOLLO SCIENTIFIC

OR46341-100G OR46341-25G OR46341-5G
77409-924EA 144.68 USD
77409-924 77409-926 77409-928
Tricyclohexylphosphine 97%
Tricyclohexylphosphine
Formula: C₁₈H₃₃P
Storage Temperature: Ambient
MDL Number: MFCD00003853
CAS Number: 2622-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR