(3R)-(+)-3-(Dimethylamino)pyrrolidine 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: (R)-(+)-3-(Dimethylamino)pyrrolidine, (R)-N,N-Dimethylpyrrolidin-3-amine

OR4630-1G
77409-922EA 72.34 USD
77409-922
(3R)-(+)-3-(Dimethylamino)pyrrolidine 97%
(3R)-(+)-3-(Dimethylamino)pyrrolidine
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 166 °C
Density: 0.899 g/cm³ (25 °C)
Flash Pt: 51.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00191347
CAS Number: 132958-72-6
UN: 2734
ADR: 8+3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR