3-Hydroxy-p-anisaldehyde
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde
OR28774-100G
OR28774-1KG
OR28774-500G
77408-216EA
110.04
USD
77408-216
77408-218
77408-220
3-Hydroxy-p-anisaldehyde
3-Hydroxy-p-anisaldehyde
Formula:
C₈H₈O₃ Boiling Pt: 179 °C/15 mmHg Melting Pt: 112…116 °C Density: 1.2 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003369 CAS Number: 621-59-0 EINECS: 210-694-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...