3-Hydroxy-p-anisaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde

OR28774-100G OR28774-1KG OR28774-500G
77408-216EA 110.04 USD
77408-216 77408-218 77408-220
3-Hydroxy-p-anisaldehyde
3-Hydroxy-p-anisaldehyde
Formula: C₈H₈O₃
Boiling Pt: 179 °C/15 mmHg
Melting Pt: 112…116 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR