Orotic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR28621-100G
77408-126EA 54.74 USD
77408-126
Orotic acid
Orotic acid
Formula: C₅H₄N₂O₄
Melting Pt: 300…347 °C
Storage Temperature: Ambient
MDL Number: MFCD00006027
CAS Number: 65-86-1
EINECS: 200-619-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR