6-Chloro-m-toluidine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR2643-1G
77407-788EA 21.26 USD
77407-788
6-Chloro-m-toluidine 97%
6-Chloro-m-toluidine
Formula: C₇H₈ClN
Boiling Pt: 228…230 °C
Melting Pt: 29…30 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00007674
CAS Number: 95-81-8
EINECS: 202-454-7
UN: 2239
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR