1,8-Octanediol 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1,8-Dihydroxy octane
OR2431-100G
OR2431-1KG
OR2431-500G
77407-466EA
63.16
USD
77407-466
77407-468
77407-470
1,8-Octanediol 98%
1,8-Octanediol
Formula:
C₈H₁₈O₂ Boiling Pt: 71…73 °C/20 mmHg Melting Pt: 59…61 °C Flash Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002989 CAS Number: 629-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...