Benzo[b]thiophene-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Benzothiophene-3-carboxylic acid

OR23171-5G OR23171-1G OR23171-250MG
77407-154EA 186.44 USD
77407-154 77407-150 77407-152
Benzo[b]thiophene-3-carboxylic acid
Benzo[b]thiophene-3-carboxylic acid
Formula: C₉H₆O₂S
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD01846406
CAS Number: 5381-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR