2-Bromopyridine
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR23057-100G
OR23057-1KG
OR23057-50G
77407-050EA
34.64
USD
77407-050
77407-052
77407-054
2-Bromopyridine
2-Bromopyridine
Formula:
C₅H₄BrN Boiling Pt: 192…194 °C Density: 1.657 g/cm³ (25 °C) Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006219 CAS Number: 109-04-6 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...