2-Indolinone
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone
OR30278-25G
OR30278-5G
77406-896EA
23.28
USD
77406-896
77406-898
2-Indolinone
2-Indolinone
Formula:
C₈H₇NO MW: 133.15 g/mol Boiling Pt: 313 Melting Pt: 123…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005711 CAS Number: 59-48-3 EINECS: 200-429-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...