2-Methyl-N-[2-(pyridin-2-yl)ethyl]aniline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-{2-[(2-Methylphenyl)amino]ethyl}pyridine, 2-{[2-(Pyridin-2-yl)ethyl]amino}toluene

OR21161-100MG
77406-632EA 48.9 USD
77406-632
2-Methyl-N-[2-(pyridin-2-yl)ethyl]aniline
2-Methyl-N-[2-(pyridin-2-yl)ethyl]aniline
Formula: C₁₄H₁₆N₂
MW: 212.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023534
CAS Number: 92733-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR