Ethyl-2-(bromomethyl)acrylate 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR17520-25G OR17520-5G
77406-016EA 79.46 USD
77406-016 77406-018
Ethyl-2-(bromomethyl)acrylate 95%
Ethyl-2-(bromomethyl)acrylate
Formula: C₆H₉BrO₂
Boiling Pt: 38 °C/0.8 mmHg
Density: 1.398 g/cm³ (25 °C)
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00031518
CAS Number: 17435-72-2
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR