Iodoethane 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR16933-100G
OR16933-500G
77405-850EA
83.54
USD
77405-850
77405-852
Iodoethane 98%
Iodoethane
Formula:
C₂H₅I Boiling Pt: 69…73 °C Melting Pt: –108 °C Density: 1.95 g/cm³ (25 °C) Flash Pt: 53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001091 CAS Number: 75-03-6 EINECS: 200-833-1 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...