Morpholine 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR13977-100G
77404-776EA 21.26 USD
77404-776
Morpholine 99%
Morpholine
Formula: C₄H₉NO
Boiling Pt: 129 °C
Melting Pt: –7…–5 °C
Density: 0.996 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00005972
CAS Number: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8+3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR