2,2':6',2''-Terpyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: TERPY

OR13730-1G
77404-726EA 101.88 USD
77404-726
2,2':6',2''-Terpyridine 97%
2,2':6',2''-Terpyridine
Formula: C₁₅H₁₁N₃
MW: 233.27 g/mol
Boiling Pt: 370 °C
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006213
CAS Number: 1148-79-4
EINECS: 214-559-5
UN: 2811
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR