1,3-Dibromobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR13132-100G OR13132-25G
77404-540EA 28.52 USD
77404-540 77404-542
1,3-Dibromobenzene 98%
1,3-Dibromobenzene
Formula: C₆H₄Br₂
Boiling Pt: 218…220 °C
Melting Pt: –7 °C
Density: 1.952 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000078
CAS Number: 108-36-1
EINECS: 203-574-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR