(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methylamine 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR12318-1G
77404-268EA 212.94 USD
77404-268
(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methylamine 95%
(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methylamine
Formula: C₁₁H₁₂N₂S
Storage Temperature: Refrigerator
MDL Number: MFCD05865105
CAS Number: 165735-95-5
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR