2-Bromethylamine hydrobromide 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

OR1160-100G OR1160-1KG OR1160-500G
77403-970EA 21.36 USD
77403-970 77403-972 77403-974
2-Bromethylamine hydrobromide 98%
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR